missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-2-Cyclohexylglycine, 95%

Catalog No. AAH6338106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6338106 5 g
AAH6338103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6338106 Supplier Thermo Scientific Chemicals Supplier No. H6338106
Only null left

CAS: 14328-52-0 | C8H15NO2 | 157.213 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14328-52-0
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD01311678
InChI Key WAMWSIDTKSNDCU-SSDOTTSWSA-N
Synonym d-cyclohexylglycine, d-alpha-cyclohexylglycine, r-2-amino-2-cyclohexylacetic acid, h-d-chg-oh, 2-cyclohexyl-d-glycine, 2r-amino cyclohexyl acetic acid, 2r-2-amino-2-cyclohexylacetic acid, d-2-cyclohexylglycine, d-cyclohexyl glycine, h-cyclohexyl-d-gly-oh
PubChem CID 736849
IUPAC Name (2R)-2-amino-2-cyclohexylacetic acid
SMILES C1CCC(CC1)C(C(=O)O)N

Specifications

Beilstein 3196806
Quantity 5 g
Formula Weight 157.21
Percent Purity 95%
Physical Form Solid
Chemical Name or Material D-(-)-2-Cyclohexylglycine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only