missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-2-Aminobutyric acid, 98+%

Catalog No. AAL1409603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1409603 1 g
AAL1409606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1409603 Supplier Thermo Scientific Chemicals Supplier No. L1409603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2623-91-8 | C4H9NO2 | 103.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2623-91-8
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
MDL Number MFCD00064414
InChI Key QWCKQJZIFLGMSD-GSVOUGTGSA-N
Synonym d-2-aminobutyric acid, r-2-aminobutanoic acid, d---2-aminobutyric acid, 2r-2-aminobutanoic acid, d-alpha-aminobutyric acid, r---2-aminobutyric acid, h-d-abu-oh, d--2-aminobutyric acid, d-2-aminobutyrate, d-2-aminobutanoic acid
PubChem CID 439691
ChEBI CHEBI:28797
IUPAC Name (2R)-2-aminobutanoic acid
SMILES CC[C@@H](N)C(O)=O

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 1720934
Optical Rotation −20.5° (c=4.8 in 6N HCl)
Formula Weight 103.12
Percent Purity ≥98%
Chemical Name or Material D-(-)-2-Aminobutyric acid
EINECSNumber 220-084-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only