missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(-)-2-(2,5-Dihydrophenyl)glycine 97.0+%, TCI America™
$452.97 - $452.97
Chemical Identifiers
| CAS | 26774-88-9 |
|---|---|
| Molecular Formula | C8H11NO2 |
| Molecular Weight (g/mol) | 153.181 |
| MDL Number | MFCD00137746 |
| InChI Key | JBJJTCGQCRGNOL-SSDOTTSWSA-N |
| Synonym | r-2-amino-2-cyclohexa-1,4-dien-1-yl acetic acid, r---2-2,5-dihydrophenyl glycine, unii-451nq27tg5, 2,5-dihydrophenylglycine, d, r---2-amino-2-cyclohexa-1,4-dienyl acetic acid, d---2-2,5-dihydrophenyl glycine, r-alpha-amino-1,4-cyclohexadiene-1-acetic acid, 2r-2-amino-2-cyclohexa-1,4-dienylacetic acid, r-amino cyclohexa-1,4-dien-1-yl acetic acid, d-cyclohexadienylglycine |
| PubChem CID | 1268102 |
| IUPAC Name | (2R)-2-amino-2-cyclohexa-1,4-dien-1-ylacetic acid |
| SMILES | C1C=CCC(=C1)C(C(=O)O)N |
Chemical Identifiers
| 26774-88-9 | |
| 153.181 | |
| JBJJTCGQCRGNOL-SSDOTTSWSA-N | |
| 1268102 | |
| C1C=CCC(=C1)C(C(=O)O)N |
| C8H11NO2 | |
| MFCD00137746 | |
| r-2-amino-2-cyclohexa-1,4-dien-1-yl acetic acid, r---2-2,5-dihydrophenyl glycine, unii-451nq27tg5, 2,5-dihydrophenylglycine, d, r---2-amino-2-cyclohexa-1,4-dienyl acetic acid, d---2-2,5-dihydrophenyl glycine, r-alpha-amino-1,4-cyclohexadiene-1-acetic acid, 2r-2-amino-2-cyclohexa-1,4-dienylacetic acid, r-amino cyclohexa-1,4-dien-1-yl acetic acid, d-cyclohexadienylglycine | |
| (2R)-2-amino-2-cyclohexa-1,4-dien-1-ylacetic acid |
Specifications
| 26774-88-9 | |
| MFCD00137746 | |
| r-2-amino-2-cyclohexa-1,4-dien-1-yl acetic acid, r---2-2,5-dihydrophenyl glycine, unii-451nq27tg5, 2,5-dihydrophenylglycine, d, r---2-amino-2-cyclohexa-1,4-dienyl acetic acid, d---2-2,5-dihydrophenyl glycine, r-alpha-amino-1,4-cyclohexadiene-1-acetic acid, 2r-2-amino-2-cyclohexa-1,4-dienylacetic acid, r-amino cyclohexa-1,4-dien-1-yl acetic acid, d-cyclohexadienylglycine | |
| C1C=CCC(=C1)C(C(=O)O)N | |
| 153.181 | |
| 153.18 | |
| Crystalline Powder |
| C8H11NO2 | |
| 5 g | |
| JBJJTCGQCRGNOL-SSDOTTSWSA-N | |
| (2R)-2-amino-2-cyclohexa-1,4-dien-1-ylacetic acid | |
| 1268102 | |
| ≥97.0% (T) | |
| D-(-)-2-(2,5-Dihydrophenyl)glycine |
Safety and Handling
TSCA : No