missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-2-(2,5-Dihydrophenyl)glycine 97.0+%, TCI America™

Catalog No. D25395G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D25395G Supplier TCI America Supplier No. D25395G
Only null left

Chemical Identifiers

CAS 26774-88-9
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.181
MDL Number MFCD00137746
InChI Key JBJJTCGQCRGNOL-SSDOTTSWSA-N
Synonym r-2-amino-2-cyclohexa-1,4-dien-1-yl acetic acid, r---2-2,5-dihydrophenyl glycine, unii-451nq27tg5, 2,5-dihydrophenylglycine, d, r---2-amino-2-cyclohexa-1,4-dienyl acetic acid, d---2-2,5-dihydrophenyl glycine, r-alpha-amino-1,4-cyclohexadiene-1-acetic acid, 2r-2-amino-2-cyclohexa-1,4-dienylacetic acid, r-amino cyclohexa-1,4-dien-1-yl acetic acid, d-cyclohexadienylglycine
PubChem CID 1268102
IUPAC Name (2R)-2-amino-2-cyclohexa-1,4-dien-1-ylacetic acid
SMILES C1C=CCC(=C1)C(C(=O)O)N

Specifications

Quantity 5 g
Formula Weight 153.18
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-(-)-2-(2,5-Dihydrophenyl)glycine