missing translation for 'onlineSavingsMsg'
Learn More

Promega CytoTox-Glo™ Cytotoxicity Assay

Catalog No. PRG9291
Encompass
Change view
Click to view available options
Quantity:
10 mL
2 x 50 mL
5 x 10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG9291 5 x 10 mL
PRG9292 2 x 50 mL
PRG9290 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRG9291 Supplier Promega Supplier No. G9291
Only null left

The CytoTox-Glo™ Assay is a luminescent cytotoxicity assay that measures the relative number of dead cells in a population.

  • Measures Cytotoxicity Due to Loss of Membrane Integrity
  • Measures extracellular activity of dead-cell protease
  • Easy and fast add-mix-measure protocol
  • Powerful luminescent detection

Specifications

Quantity 5 x 10 mL
Content And Storage -30°C to -10°C