Learn More
Promega CytoTox-Glo™ Cytotoxicity Assay
The CytoTox-Glo™ Assay is a luminescent cytotoxicity assay that measures the relative number of dead cells in a population.
Supplier:  Promega G9290
Encompass
 View more versions of this product
Catalog No. PRG9290
Only null left
Description
Description
  • Measures Cytotoxicity Due to Loss of Membrane Integrity
  • Measures extracellular activity of dead-cell protease
  • Easy and fast add-mix-measure protocol
  • Powerful luminescent detection
Specifications
Chemical Identifiers
-30°C to -10°C
Specifications
10 mL
-30°C to -10°C
Product Suggestions
Product Suggestions