Learn More
Epredia™ Cytospin™ 4 Cytocentrifuge Sealed Head Rotor
Catalog No. SH59910019
Change view
Click to view available options
Product Type:
Head Seals
Sealed Head Rotor
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
SH59910019 Head Seals
59910018 Sealed Head Rotor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH59910019 Supplier Epredia™ Supplier No. 59910019
Use these accessories with the Epredia™ Cytospin™ 4 Cytocentrifuge.
  • Shorten preparation time by having a second sealed head ready when the first finishes processing
  • Autoclavable
  • 12-chamber head seals completely
Specifications
For Use With (Equipment) Cytospin 4 Cytocentrifuge
Product Type Head Seals