missing translation for 'onlineSavingsMsg'
Learn More

Cytosine 98.0+%, TCI America™

Catalog No. p-7124489
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C05285G 5 g
C052825G 25 g
C0528100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Catalog No. C05285G missing translation for 'mfr' TCI America missing translation for 'supplierNo' C05285G

Chemical Identifiers

CAS 71-30-7
Molecular Formula C4H5N3O
Molecular Weight (g/mol) 111.10
MDL Number MFCD00006034
InChI Key OPTASPLRGRRNAP-UHFFFAOYSA-N
Synonym cytosine, 4-amino-2-hydroxypyrimidine, cytosinimine, 2 1h-pyrimidinone, 4-amino, 4-amino-2 1h-pyrimidinone, 4-aminopyrimidin-2 1h-one, 6-aminopyrimidin-2 1h-one, 2 1h-pyrimidinone, 6-amino, cyt, 4-amino-2-oxo-1,2-dihydropyrimidine
PubChem CID 597
ChEBI CHEBI:16040
IUPAC Name 6-amino-1,2-dihydropyrimidin-2-one
SMILES NC1=CC=NC(=O)N1

Specifications

Color White
Quantity 5 g
Formula Weight 111.10
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cytosine
EINECSNumber (9)-0509
RTECSNumber UW7350150
TSCA Yes