missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Cytosine-beta-D-arabinofuranose hydrochloride
Interferes with the synthesis of DNA | C9H14ClN3O5 | 279.68 g/mol
Supplier: Thermo Scientific Chemicals J6567106
Description
Interferes with the synthesis of DNA
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Solid | |
| Cytosine-beta-D-arabinofuranose hydrochloride | |
| MFCD00012839 | |
| Keep cold | |
| Cl.NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O | |
| White |
| 279.68,279.68 | |
| C9H14ClN3O5 | |
| 5 g,5 g | |
| KCURWTAZOZXKSJ-JBMRGDGGSA-N | |
| 4-amino-1-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one hydrochloride |
RUO – Research Use Only