Learn More
StatLab™ CytomeGallonovirus (CMV) IHC Tissue Control Slides
CMV Tissue Control Slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
Supplier: StatLab™ CSC0925P
Description
- Use these slides with confidence to evaluate stain quality and performance in histology laboratories
- Control slides are created using Millennia adhesion white glass slides with 90 degree corners
- Each package includes specific staining protocol
- Stained slide is not included
- Tissue: Known Positive
- Special Stain or Antibody: CMV
Specifications
| Control Slide | |
| 25 Slides/Pk. |