missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- For use in Cytospin centrifuge
- 75 mm x 25 mm slides printed with circle(s) on the surface
- Well(s) placed in the exact position where the centrifuge specimen is deposited
- PK144
- 10 packs per case
Specifications
| 90°, Ground | |
| Glass | |
| Non-sterile | |
| 75 mm | |
| Clear |
| Single Ringed Cytology Slide | |
| 1 | |
| 1 mm | |
| 25 mm | |
| One Ring Cytology Slides |