missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cytochalasin D

Catalog No. PHZ1063
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PHZ1063 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PHZ1063 Supplier Invitrogen™ Supplier No. PHZ1063

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cytochalasin D is a cell permeable fungal toxin that binds to the barbed end of actin filaments inhibiting both the association and dissociation of subunits.

Cytochalasin D is a cell permeable fungal toxin that binds to the barbed end of actin filaments inhibiting both the association and dissociation of subunits. This compound causes the disruption of actin filaments and inhibition of actin polymerization.

Format/Formulation: Lyophilized

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Gene ID (Entrez) 60
Shipping Condition Dry Ice
Specific Inhibit or Target Actin
Sterility Non-sterile
Quantity 1 mg
Product Type Cytochalasin D
Inhibitor Properties Chemical
Molecular Weight (g/mol) 507.6
Inhibitors Cytoskeletal Inhibitor
Form Solid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.