Learn More
Invitrogen™ CytKick™ Max Autosampler Upgrade Kit
Enables the upgrade of a CytKick Autosampler to a CytKick Max Autosampler
Supplier:  Invitrogen™ A43045
Catalog No. A43045
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • The CytKick Max Autosampler Upgrade Kit enables the upgrade of a CytKick Autosampler to a CytKick Max Autosampler.
  • The upgrade is carried out by either a field service engineer or field application specialist and provides the following features: faster plate times (22 minutes for a 96-well plate or 88 minutes for a 384-well plate) with a boost mode, user-definable customized plate definitions allowing the use of any ANSI spec plates, evaporation protection with foil covers, automated tube sample acquisition for 1.5- and 2.0-mL Eppendorf tubes, and passive cooling capability.
Specifications
Specifications
CytKick
Room Temperature
Autosampler Upgrade Kit
1 kit
Attune Acoustic Focusing Cytometer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.