missing translation for 'onlineSavingsMsg'
Learn More

Cytidine 5'-Monophosphate 98.0+%, TCI America™

Catalog No. p-7125121
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16751G 1 g
C16755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16751G Supplier TCI America Supplier No. C16751G

Chemical Identifiers

CAS 63-37-6
Molecular Formula C9H14N3O8P
Molecular Weight (g/mol) 323.20
MDL Number MFCD00006544
InChI Key IERHLVCPSMICTF-NHBCCFCANA-N
Synonym 5'-cytidylic acid, cytidylic acid, cytidine 5'-monophosphate, cytidine monophosphate, cytidine-5'-monophosphate, 5'-cmp, cytidylate, cytidine 5'-phosphate, cmp nucleotide, cytidine 5'-phosphoric acid
PubChem CID 6131
ChEBI CHEBI:17361
IUPAC Name {[(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}phosphonic acid
SMILES NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP(O)(O)=O)[C@@H](O)[C@H]1O

Specifications

Color White
Quantity 1 g
Formula Weight 323.20
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cytidine 5′-Monophosphate
EINECSNumber (8)-0281
RTECSNumber HA3980000
TSCA Yes
Recommended Storage Refrigerator