missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Cytidine 5'-monophosphate hydrate, 99%
Chemical Identifiers
| CAS | 63-37-6 |
|---|---|
| Molecular Formula | C9H14N3O8P·xH2O |
| Molecular Weight (g/mol) | 323.2 |
| MDL Number | MFCD00149426 |
| InChI Key | IERHLVCPSMICTF-XVFCMESISA-N |
| Synonym | 5'-cytidylic acid, cytidylic acid, cytidine 5'-monophosphate, cytidine monophosphate, cytidine-5'-monophosphate, 5'-cmp, cytidylate, cytidine 5'-phosphate, cmp nucleotide, cytidine 5'-phosphoric acid |
| PubChem CID | 6131 |
| ChEBI | CHEBI:17361 |
| IUPAC Name | [(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate |
| SMILES | C1=CN(C(=O)N=C1N)C2C(C(C(O2)COP(=O)(O)O)O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC226210010
|
Thermo Scientific™
226210010 |
1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC226210050
|
Thermo Scientific™
226210050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 63-37-6 | |
| 323.2 | |
| IERHLVCPSMICTF-XVFCMESISA-N | |
| 6131 | |
| [(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate |
| C9H14N3O8P·xH2O | |
| MFCD00149426 | |
| 5'-cytidylic acid, cytidylic acid, cytidine 5'-monophosphate, cytidine monophosphate, cytidine-5'-monophosphate, 5'-cmp, cytidylate, cytidine 5'-phosphate, cmp nucleotide, cytidine 5'-phosphoric acid | |
| CHEBI:17361 | |
| C1=CN(C(=O)N=C1N)C2C(C(C(O2)COP(=O)(O)O)O)O |
Specifications
| 63-37-6 | |
| 5% max. (105°C, 5 hrs) | |
| 98.5% min. (HPLC) | |
| C9H14N3O8P·xH2O | |
| 1g | |
| Solubility in water: soluble | |
| IERHLVCPSMICTF-XVFCMESISA-N | |
| [(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate | |
| 6131 | |
| 323.2 | |
| Cytidine 5′-monophosphate hydrate |
| 0.41 to 0.49 (A250/A260), 2.03 to 2.17 (A280/A260) | |
| Authentic | |
| Glass bottle | |
| MFCD00149426 | |
| 5'-cytidylic acid, cytidylic acid, cytidine 5'-monophosphate, cytidine monophosphate, cytidine-5'-monophosphate, 5'-cmp, cytidylate, cytidine 5'-phosphate, cmp nucleotide, cytidine 5'-phosphoric acid | |
| 98 to 102% (lambda max. 280nm,pH 2) molar absorptivity 13000+/-2%,on dry basis | |
| C1=CN(C(=O)N=C1N)C2C(C(C(O2)COP(=O)(O)O)O)O | |
| 323.2 | |
| CHEBI:17361 | |
| 99% |
RUO â Research Use Only