missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cytidine 5'-monophosphate hydrate, 99%

Catalog No. AC226210010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC226210010 1 g Glass bottle
AC226210050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC226210010 Supplier Thermo Scientific Chemicals Supplier No. 226210010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 63-37-6 | C9H14N3O8P | 323.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63-37-6
Molecular Formula C9H14N3O8P
Molecular Weight (g/mol) 323.20
MDL Number MFCD00006544
InChI Key IERHLVCPSMICTF-NHBCCFCANA-N
Synonym 5'-cytidylic acid, cytidylic acid, cytidine 5'-monophosphate, cytidine monophosphate, cytidine-5'-monophosphate, 5'-cmp, cytidylate, cytidine 5'-phosphate, cmp nucleotide, cytidine 5'-phosphoric acid
PubChem CID 6131
ChEBI CHEBI:17361
IUPAC Name [(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
SMILES NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP(O)(O)=O)[C@@H](O)[C@H]1O

Specifications

Absorbance 0.41 to 0.49 (A250/A260), 2.03 to 2.17 (A280/A260)
Melting Point 222.0°C
Color White
Loss on Drying 5% max. (105°C, 5 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble
Spectrophotometry 98 to 102% (lambda max. 280nm,pH 2) molar absorptivity 13000+/-2%,on dry basis
Formula Weight 323.2
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Cytidine 5′-monophosphate hydrate
Show More Show Less

RUO – Research Use Only