Learn More
Agilent BioTek Cytation 5 Cell Imaging Multi-Mode Reader

Cytation 5 combines automated microscopy and conventional microplate detection in a configurable, upgradable platform. For Research Use Only. Not for use in diagnostic procedures.
Supplier: Agilent BioTek CYT5MFWSN
Description
Cytation 5 combines automated microscopy and conventional microplate detection in a configurable, upgradable platform. The microscopy module offers up to 60x magnification in fluorescence, brightfield, high contrast brightfield, color brightfield, and phase contrast to address many applications and workflows
The multi-mode detection modules include filter- and monochromator-based fluorescence detection, luminescence, and UV-Vis absorbance detection. Gen5 software provides complete control over all imaging and data capture, plus powerful image and data analysis.
Chemical Identifiers
| Cell Imaging Multi-Mode Microplate Reader | |
| Sony CMOS, 16-bit Gray Scale, Standard or WFOV | |
| 230 to 999 nm, 1 nm increments | |
| Absorbance: 0 to 4 OD; Fluorescence Intensity: 7 decades; Luminesence: >6 decades | |
| Fluorescence, phase contrast, brightfield, high contrast brightfield, color brightfield and phase contrast | |
| 2 Syringe Pumps | |
| Xenon Flash; 100 mW 680 nm laser (Alpha Detection) | |
| 4-Zone incubation to 65°C with Condensation Control (±0.2°C at 37°C) | |
| 51.4 cm | |
| 44.5 cm | |
| 36.3 kg | |
| 41.6 cm | |
| 250 W max. |
| Linear, Orbital, Double Orbital | |
| 0 to 20% | |
| Monochromator: ±2 nm; Dispenser: ±1 μL or 2% | |
| BioStack and Third Party Automation Compatible; BioSpa 8 Automated Incubator Compatible | |
| 5 to 1000 μL, 1 μL increments | |
| End point, kinetic, spectral scanning and well area scanning | |
| 1.25x, 2.5x (2.25x eff), 2.5x (2.75x eff), 4x, 10x, 20x, 40x, 60x | |
| 20.2 in. | |
| 17.5 in. | |
| 80 lb. | |
| 16.4 in. | |
| CE and TUV marked. RoHS compliant. |
Specifications
| Cell Imaging Multi-Mode Microplate Reader | |
| Sony CMOS, 16-bit Gray Scale, Standard or WFOV | |
| 230 to 999 nm, 1 nm increments | |
| Absorbance: 0 to 4 OD; Fluorescence Intensity: 7 decades; Luminesence: >6 decades | |
| Fluorescence, phase contrast, brightfield, high contrast brightfield, color brightfield and phase contrast | |
| Wide FOV camera; Fluorescence/brightfield/color brightfield imaging; Filter-based fluorescence intensity/fluorescence polarization/time resolved fluorescence/luminescence; Monochromator-based UV-Vis absorbance/fluorescence intensity/time resolved fluorescence (secondary)/luminescence | |
| 2 Syringe Pumps | |
| End point, kinetic, spectral scanning and well area scanning | |
| 1.25x, 2.5x (2.25x eff), 2.5x (2.75x eff), 4x, 10x, 20x, 40x, 60x | |
| 4-Zone incubation to 65°C with Condensation Control (±0.2°C at 37°C) | |
| 51.4 cm | |
| 44.5 cm | |
| 36.3 kg | |
| 41.6 cm | |
| 250 W max. |
| Linear, Orbital, Double Orbital | |
| 0 to 20% | |
| Monochromator: ±2 nm; Dispenser: ±1 μL or 2% | |
| BioStack and Third Party Automation Compatible; BioSpa 8 Automated Incubator Compatible | |
| 5 to 1000 μL, 1 μL increments | |
| CYT5MFW | |
| Monochromator: 6- to 384-well plates; Filters: 6-to 1536 well plates; Imaging: 6-to 1536 well plates; Take3 Plate compatible | |
| Xenon Flash; 100 mW 680 nm laser (Alpha Detection) | |
| Gen5 Microplate Reader and Imager Software included; Gen5 Image+ and Image Prime software available for full image analysis; Gen5 Secure for 21 CFR Part 11 compliance (option) | |
| 20.2 in. | |
| 17.5 in. | |
| 80 lb. | |
| 16.4 in. | |
| CE and TUV marked. RoHS compliant. |