Learn More
Agilent BioTek Cytation 5 Cell Imaging Multi-Mode Reader
Catalog No. BTCYT5FAV
Change view
Click to view available options
Model:
CYT5MPW
CYT5MV
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
BTCYT5FAV -
BTCYT5FW -
BTCYT5FAW -
BTCYT5MPW CYT5MPW
BTCYT5MV CYT5MV
BTCYT5MFV -
BTCYT5MFAV -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. BTCYT5FAV Supplier Agilent BioTek Supplier No. CYT5FAVSN
Cytation 5 combines automated microscopy and conventional microplate detection in a configurable, upgradable platform. For Research Use Only. Not for use in diagnostic procedures.
Cytation 5 combines automated microscopy and conventional microplate detection in a configurable, upgradable platform. The microscopy module offers up to 60x magnification in fluorescence, brightfield, high contrast brightfield, color brightfield, and phase contrast to address many applications and workflows
The multi-mode detection modules include filter- and monochromator-based fluorescence detection, luminescence, and UV-Vis absorbance detection. Gen5 software provides complete control over all imaging and data capture, plus powerful image and data analysis.