Learn More
MilliporeSigma™ Cysteine Sulfenic Acid Probe (DCP-Bio1)
Catalog No. NS12265MG
Change view
Click to view available options
Quantity:
5 x 1 mg
Catalog No. Quantity
NS12265MG 5 x 1 mg
1 options
Catalog No. NS12265MG Supplier MilliporeSigma™ Supplier No. NS12265MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
DCP-Bio1: 1mg dry powder, stored under argon
DCP-Bio1, which consists of the sulfenic acid-reactive 3-(2,4-dioxocyclohexyl) propyl (DCP) appended to biotin, is a useful reagent for characterizing cellular cysteine sulfenic acid
  • New reagents have been developed to simplify the analysis and isolation of proteins bearing cysteine sulfenic acid
  • DCP-Bio1, which consists of the sulfenic acid-reactive 3-(2,4-dioxocyclohexyl) propyl (DCP) appended to biotin, is a useful reagent for characterizing cellular cysteine sulfenic acid 
Specifications
Content And Storage -20°C
Quantity 5 x 1 mg