missing translation for 'onlineSavingsMsg'
Learn More

Cyromazine 98.0+%, TCI America™

Catalog No. c236625g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
C236625G 25 g
C23665G 5 g
2 options

Catalog No. C236625G

Supplier: TCI America C236625G

Only null left

Chemical Identifiers

CAS 66215-27-8
Molecular Formula C6H10N6
Molecular Weight (g/mol) 166.188
MDL Number MFCD00078650
InChI Key LVQDKIWDGQRHTE-UHFFFAOYSA-N
Synonym cyromazine, larvadex, vetrazin, trigard, cyclopropylmelamine, cyromazin, vetrazine, neporex, citation, cypromazine
PubChem CID 47866
ChEBI CHEBI:30260
IUPAC Name 2-N-cyclopropyl-1,3,5-triazine-2,4,6-triamine
SMILES C1CC1NC2=NC(=NC(=N2)N)N

Specifications

Melting Point 224°C
Color White-Yellow
Quantity 25 g
Formula Weight 166.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cyromazine
RTECSNumber XZ1056500
TSCA No