missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyproconazole
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
Cyproconazole is used to inhibit demeythylation in fungal sterol synthesis. And it is used on greenhouse and field-grown roses and as a wood preservative, surface application or pressure treatment of wood for above-ground uses, including siding, plywood, millwork, shingles, lumber.
Solubility
Soluble in acetone, ethanol, xylene, and dimethyl sulfoxide.
Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
Cyproconazole is used to inhibit demeythylation in fungal sterol synthesis. And it is used on greenhouse and field-grown roses and as a wood preservative, surface application or pressure treatment of wood for above-ground uses, including siding, plywood, millwork, shingles, lumber.
Solubility
Soluble in acetone, ethanol, xylene, and dimethyl sulfoxide.
Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Type | Cyproconazole |
| Physical Form | Crystalline |
| Melting Point | 108°C |
| Color | Brown |
| Quantity | 5 g |
| Flash Point | >100°C (212°F) |
| UN Number | UN3077 |
| Boiling Point | >250°C |
| Beilstein | 8396421 |
| Merck Index | 14,2771 |
| Show More |
Safety and Handling
| Hazard Category | H301-H360D-H373 |
| Hazard Statement | GHS H Statement H361-H302 Suspected of damaging fertility or the unborn child. Harmful if swallowed. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P281-P301+P310-P308+P313-P330-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| RTECSNumber | XZ4803250 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only