missing translation for 'onlineSavingsMsg'
Learn More

Cyproconazole

Catalog No. AAJ6314206
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6314206 5 g
AAJ6314209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6314206 Supplier Thermo Scientific Chemicals Supplier No. J6314206

Triazole fungicide which causes hepatocellular adenomas and carcinomas | CAS: 94361-06-5 | C15H18ClN3O | 291.779 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Cyproconazole is used to inhibit demeythylation in fungal sterol synthesis. And it is used on greenhouse and field-grown roses and as a wood preservative, surface application or pressure treatment of wood for above-ground uses, including siding, plywood, millwork, shingles, lumber.

Solubility
Soluble in acetone, ethanol, xylene, and dimethyl sulfoxide.

Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 94361-06-5
Molecular Formula C15H18ClN3O
Molecular Weight (g/mol) 291.779
MDL Number MFCD01678672
InChI Key UFNOUKDBUJZYDE-UHFFFAOYSA-N
PubChem CID 86132
ChEBI CHEBI:83748
IUPAC Name 2-(4-chlorophenyl)-3-cyclopropyl-1-(1,2,4-triazol-1-yl)butan-2-ol
SMILES CC(C1CC1)C(CN2C=NC=N2)(C3=CC=C(C=C3)Cl)O

Specifications

Type Cyproconazole
Physical Form Crystalline
Melting Point 108°C
Color Brown
Quantity 5 g
Flash Point >100°C (212°F)
UN Number UN3077
Boiling Point >250°C
Beilstein 8396421
Merck Index 14,2771
Solubility Information Soluble in acetone,ethanol,xylene,and dimethyl sulfoxide.
Formula Weight 291.78
Chemical Name or Material Cyproconazole
Show More Show Less
Hazard Category H301-H360D-H373
Hazard Statement GHS H Statement
H361-H302
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Precautionary Statement P201-P202-P260-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECSNumber XZ4803250
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only