MW 285.39 g/mol, Purity >98%. First selective small conductance Ca+-activated K+ channel (SK) activator. Selectively opens SK2 and SK3 channels (EC values are 5.6 and 14 M, respectively). Shows no activity at SK1 and IK. Additionally reduces activity and excitability of dopamine neurons. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement