missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Cylindrical Alnico Magnet Pair with Keepers

Catalog No. S27738
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S27738 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27738 Supplier Eisco™ Supplier No. PH0793B

An excellent tool for visualizing and kinesthetically experiencing the push and pull of magnetic fields. The magnet keeper helps to preserve the magnet's strength by completing the magnetic circuit and prohibiting external metal from being attracted to the magnet.

  • Includes 2 cylindrical magnets, painted red
  • Pink dot denotes opposing poles
  • Keepers at the end of the magnets maintain magnet's strength
  • Supplied with a box for long term storage

Specifications

Grade 6 to Undergraduate
For Use With (Application) Experiencing The Push and Pull of Magnetic Fields
Includes 2 Magnets, 2 Magnet Keepers
Material Alnico (Aluminum, Nickel, Cobalt, and Iron)
Quantity 2 Pk.
Length (English) 2 in.
Length (Metric) 50.8 mm
Class Size 1 Lab Group
Product Type Cylindrical Magnet