missing translation for 'onlineSavingsMsg'
Learn More

Cyclosporin A 97.0+%, TCI America™

Catalog No. C24081G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24081G 1 g
C2408100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24081G Supplier TCI America Supplier No. C24081G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 59865-13-3
Molecular Formula C62H111N11O12
Molecular Weight (g/mol) 1202.64
MDL Number MFCD00274558
InChI Key PMATZTZNYRCHOR-IMVLJIQENA-N
Synonym cyclosporin a
PubChem CID 132274082
IUPAC Name 30-ethyl-33-[(4E)-1-hydroxy-2-methylhex-4-en-1-yl]-1,4,7,10,12,15,19,25,28-nonamethyl-6,9,18,24-tetrakis(2-methylpropyl)-3,21-bis(propan-2-yl)-1,4,7,10,13,16,19,22,25,28,31-undecaazacyclotritriacontan-2,5,8,11,14,17,20,23,26,29,32-undecone
SMILES CCC1NC(=O)C(C(O)C(C)C\C=C\C)N(C)C(=O)C(C(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(C)NC(=O)C(C)NC(=O)C(CC(C)C)N(C)C(=O)C(NC(=O)C(CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C

Specifications

Melting Point 151°C
Color White
Quantity 1 g
Formula Weight 1202.64
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Cyclosporin A
RTECSNumber GZ4120000
TSCA No
Recommended Storage Refrigerator