missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cyclosporin A, 98%

Catalog No. AC457970050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457970050 5 g
AC457970010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC457970050 Supplier Thermo Scientific Chemicals Supplier No. 457970050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cyclosporin A, 98%, CAS # 59865-13-3, is a compound that shows immunosuppressive, antifungal, antirheumatic, and dermatologic activities. It is also a calcineurin and enzyme inhibitor. | CAS: 59865-13-3 | C62H111N11O12 | 1202.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Cyclosporin A is a non-polar cyclic oligopeptide produced by the fungus Tolypocladium inflatum

Chemical Identifiers

CAS 59865-13-3
Molecular Formula C62H111N11O12
Molecular Weight (g/mol) 1202.64
MDL Number MFCD00274558
InChI Key PMATZTZNYRCHOR-IMVLJIQENA-N
PubChem CID 132274082
SMILES CCC1NC(=O)C(C(O)C(C)C\C=C\C)N(C)C(=O)C(C(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(C)NC(=O)C(C)NC(=O)C(CC(C)C)N(C)C(=O)C(NC(=O)C(CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C

Specifications

Type Cyclosporin A
Melting Point 146.0°C to 151.0°C
Infrared Spectrum Authentic
Loss on Drying 2% max. (60°C, 3 hrs, 5 mm Hg)
Quantity 5 g
IUPAC Name (3S,6R,9R,12R,15R,18S,24S,30S,33R)-30-ethyl-33-[(E,1S)-1-hydroxy-2-methylhex-4-enyl]-1,4,7,10,12,15,19,25,28-nonamethyl-6,9,18,24-tetrakis(2-methylpropyl)-3,21-di(propan-2-yl)-1,4,7,10,13,16,19,22,25,28,31-undecazacyclotritriacontane-2,5,8,11,14,17,20,23,
Formula Weight 1202.61
Percent Purity 98%
Chemical Name or Material Cyclosporin A
Which solvents can I use to dissolve cyclosporin A, 98%?

Cyclosporin A is soluble in ethanol, chloroform, methylene chloride/dichloromethane, and DMSO.

Hazard Category Danger
Hazard Statement GHS H Statement Harmful if swallowed. May cause cancer. May damage fertility. May damage the unborn child.
Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Obtain special instructions before use. Use personal protective equipment as required. IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only