missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropylmethylamine 95.0+%, TCI America™
Supplier: TCI America C234525G
Specifications
| Cyclopropylmethylamine | |
| Yellow | |
| C4H9N | |
| 25 g | |
| cyclopropylmethylamine, cyclopropanemethylamine, cyclopropanemethanamine, aminomethylcyclopropane, aminomethyl cyclopropane, 1-cyclopropylmethanamine, cyclopropane methyl amine, cyclopropylmethylamin, cyclopropanmethylamine | |
| C1CC1CN | |
| 71.123 | |
| 71.12 | |
| Liquid |
| 2516-47-4 | |
| 86°C | |
| MFCD00037147 | |
| 2733 | |
| IGSKHXTUVXSOMB-UHFFFAOYSA-N | |
| cyclopropylmethanamine | |
| 75646 | |
| ≥95.0% (GC,T) |
Chemical Identifiers
| 2516-47-4 | |
| 71.123 | |
| IGSKHXTUVXSOMB-UHFFFAOYSA-N | |
| 75646 | |
| C1CC1CN |
| C4H9N | |
| MFCD00037147 | |
| cyclopropylmethylamine, cyclopropanemethylamine, cyclopropanemethanamine, aminomethylcyclopropane, aminomethyl cyclopropane, 1-cyclopropylmethanamine, cyclopropane methyl amine, cyclopropylmethylamin, cyclopropanmethylamine | |
| cyclopropylmethanamine |
Safety and Handling
TSCA : Yes