missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylmethylamine 95.0+%, TCI America™

Catalog No. C23455G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23455G 5 g
C234525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23455G Supplier TCI America Supplier No. C23455G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2516-47-4
Molecular Formula C4H9N
Molecular Weight (g/mol) 71.123
MDL Number MFCD00037147
InChI Key IGSKHXTUVXSOMB-UHFFFAOYSA-N
Synonym cyclopropylmethylamine, cyclopropanemethylamine, cyclopropanemethanamine, aminomethylcyclopropane, aminomethyl cyclopropane, 1-cyclopropylmethanamine, cyclopropane methyl amine, cyclopropylmethylamin, cyclopropanmethylamine
PubChem CID 75646
IUPAC Name cyclopropylmethanamine
SMILES C1CC1CN

Specifications

Color Yellow
Boiling Point 86°C
Quantity 5 g
UN Number 2733
Formula Weight 71.12
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Cyclopropylmethylamine