Learn More
Cyclopropylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
$508.41 - $508.41
Chemical Identifiers
| CAS | 23719-80-4 |
|---|---|
| Molecular Formula | C3H5BrMg |
| Molecular Weight (g/mol) | 145.28 |
| MDL Number | MFCD00672094 |
| InChI Key | QUPLLJGAJVXXBW-UHFFFAOYSA-M |
| Synonym | cyclopropylmagnesium bromide, grignard reagent, cyclopropylmagnesiumbromide, magnesium, bromocyclopropyl, magnesium, bromocyclopropyl, cprmgbr, cyclopropyl magnesiumbromide, magnesium,bromocyclopropyl, bromo-cyclopropyl magnesium, bromo-cyclopropyl magnesium, cyclopropylmagnesiunn bromide |
| PubChem CID | 11116189 |
| IUPAC Name | magnesium;cyclopropane;bromide |
| SMILES | Br[Mg]C1CC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH26273AE
|
Thermo Scientific Chemicals
H26273AE |
100 mL |
Each for $508.41
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt often acts as the starting nutrient mix for dropout studies where fermentation conditions required the absence of one or more essential amino acids. For classifying Yeasts based on colonial characteristics and cell morphology.
Solubility
Soluble in water (17 mg/ml), (25°C). Soluble in organic solvents.
Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, water and moisture.
Chemical Identifiers
| 23719-80-4 | |
| 145.28 | |
| QUPLLJGAJVXXBW-UHFFFAOYSA-M | |
| 11116189 | |
| Br[Mg]C1CC1 |
| C3H5BrMg | |
| MFCD00672094 | |
| cyclopropylmagnesium bromide, grignard reagent, cyclopropylmagnesiumbromide, magnesium, bromocyclopropyl, magnesium, bromocyclopropyl, cprmgbr, cyclopropyl magnesiumbromide, magnesium,bromocyclopropyl, bromo-cyclopropyl magnesium, bromo-cyclopropyl magnesium, cyclopropylmagnesiunn bromide | |
| magnesium;cyclopropane;bromide |
Specifications
| 23719-80-4 | |
| −17°C (1°F) | |
| MFCD00672094 | |
| UN2924 | |
| cyclopropylmagnesium bromide, grignard reagent, cyclopropylmagnesiumbromide, magnesium, bromocyclopropyl, magnesium, bromocyclopropyl, cprmgbr, cyclopropyl magnesiumbromide, magnesium,bromocyclopropyl, bromo-cyclopropyl magnesium, bromo-cyclopropyl magnesium, cyclopropylmagnesiunn bromide | |
| QUPLLJGAJVXXBW-UHFFFAOYSA-M | |
| magnesium;cyclopropane;bromide | |
| 11116189 | |
| 0.5 to 0.7M in THF |
| 0.968 | |
| C3H5BrMg | |
| 100 mL | |
| Air and moisture sensitive; May form a precipitate | |
| Soluble in water (17mg/ml),(25°C). Soluble in organic solvents. | |
| Br[Mg]C1CC1 | |
| 145.28 | |
| 145.28 | |
| Cyclopropylmagnesium bromide |
Safety and Handling
GHS H Statement
H225-H314-H351-H335
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
H225-H260-H302-H314-H335-H336-H351
DOTInformation : Hazard Class: 4.3; Packaging Group: I
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only