missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylcarboxamidine hydrochloride, 97%

Catalog No. AAB2018506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2018506 5 g
AAB2018514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2018506 Supplier Thermo Scientific Chemicals Supplier No. B2018506

CAS: 57297-29-7 | C4H9ClN2 | 120.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57297-29-7
Molecular Formula C4H9ClN2
Molecular Weight (g/mol) 120.58
MDL Number MFCD00053010
InChI Key JRYOZJIRAVZGMV-UHFFFAOYSA-N
Synonym cyclopropanecarboximidamide hydrochloride, cyclopropane-1-carboximidamide hydrochloride, cyclopropanecarboxamidine hydrochloride, cyclopropylcarbamidine hydrochloride, cyclopropanecarboxamidine hcl, cyclopropyl carbamidine hydrochloride, clcyopropylcarbamidine hydrochloride, cyclopropylcarboxamidine hydrochloride, cyclopropyl carbamidine hcl, cyclopropanecarboximidamide, monohydrochloride
PubChem CID 2781916
SMILES [H+].[Cl-].NC(=N)C1CC1

Specifications

Melting Point 122°C to 123°C
Quantity 5 g
Beilstein 4507255
Sensitivity Hygroscopic
IUPAC Name hydrogen cyclopropanecarboximidamide chloride
Formula Weight 120.59
Percent Purity 97%
Chemical Name or Material Cyclopropylcarboxamidine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only