missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylboronic acid, tech. 85%

Catalog No. AAH2625403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2625403 1 g
AAH2625406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2625403 Supplier Thermo Scientific Chemicals Supplier No. H2625403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 411235-57-9 | C3H7BO2 | 85.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 411235-57-9
Molecular Formula C3H7BO2
Molecular Weight (g/mol) 85.90
MDL Number MFCD04038750
InChI Key WLVKDFJTYKELLQ-UHFFFAOYSA-N
Synonym cyclopropyl boronic acid, cyclopropylboranediol, cyclopropaneboronic acid, boronic acid, cyclopropyl, cyclopropyl-boronic acid, boronic acid, cyclopropyl-9ci, cyclopropylboronicacid, boronocyclopropane, cyclopropyboronic acid, cyclopropylboromc acid
PubChem CID 2760897
IUPAC Name cyclopropylboronic acid
SMILES OB(O)C1CC1

Specifications

Melting Point 90°C to 95°C
Quantity 1 g
Formula Weight 85.9
Percent Purity 85%
Grade Technical
Chemical Name or Material Cyclopropylboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only