missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylboronic acid pinacol ester, 96%

Catalog No. AAH2764203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2764203 1 g
AAH2764206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2764203 Supplier Thermo Scientific Chemicals Supplier No. H2764203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 126689-01-8 | C9H17BO2 | 168.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 126689-01-8
Molecular Formula C9H17BO2
Molecular Weight (g/mol) 168.04
MDL Number MFCD05663847
InChI Key XGBMQBPLWXTEPM-UHFFFAOYSA-N
Synonym cyclopropylboronic acid pinacol ester, cyclopropylboronic acid, pinacol ester, 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl cyclopropane, 1,3,2-dioxaborolane, 2-cyclopropyl-4,4,5,5-tetramethyl, 2-cyclopropyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborol, cypbe, pubchem4015, pubchem7931, acmc-209bbh
PubChem CID 2758015
IUPAC Name 2-cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1CC1

Specifications

Density 0.922
Boiling Point 146°C
Flash Point 40°C (104°F)
Quantity 1 g
UN Number UN3272
Sensitivity Moisture sensitive
Formula Weight 168.04
Percent Purity 96%
Chemical Name or Material Cyclopropylboronic acid pinacol ester
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only