missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylboronic acid pinacol ester, 95%

Catalog No. AC428590010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428590010 1 g Glass bottle
AC428590050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC428590010 Supplier Thermo Scientific Chemicals Supplier No. 428590010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 126689-01-8 | C9H17BO2 | 168.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 126689-01-8
Molecular Formula C9H17BO2
Molecular Weight (g/mol) 168.04
MDL Number MFCD05663847
InChI Key XGBMQBPLWXTEPM-UHFFFAOYSA-N
Synonym cyclopropylboronic acid pinacol ester, cyclopropylboronic acid, pinacol ester, 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl cyclopropane, 1,3,2-dioxaborolane, 2-cyclopropyl-4,4,5,5-tetramethyl, 2-cyclopropyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborol, cypbe, pubchem4015, pubchem7931, acmc-209bbh
PubChem CID 2758015
IUPAC Name 2-cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1CC1

Specifications

Density 0.9220g/mL
Boiling Point 146.0°C
Flash Point 40°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 0.922
Formula Weight 168.04
Percent Purity 95%
Chemical Name or Material Cyclopropylboronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

RUO – Research Use Only