missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylamine, 99%

Catalog No. p-4510293
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC154880100 10 mL Glass bottle
AC154880500 50 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC154880100 Supplier Thermo Scientific Chemicals Supplier No. 154880100
Only null left

CAS: 765-30-0 | C3H7N | 57.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 765-30-0
Molecular Formula C3H7N
Molecular Weight (g/mol) 57.09
InChI Key HTJDQJBWANPRPF-UHFFFAOYSA-N
Synonym cyclopropylamine, aminocyclopropane, cyclopropyl amine, unii-8pr8xth1x1, 8pr8xth1x1, cyclopropanamin, cyclopropyamine, cyclopropylamin, cylopropylamine, cycloproplyamine
PubChem CID 69828
ChEBI CHEBI:34660
IUPAC Name cyclopropanamine
SMILES C1CC1N

Specifications

Melting Point -50.0°C
Color Colorless
Density 0.8240g/mL
Boiling Point 49.0°C to 50.0°C
Flash Point −25°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4190 to 1.4220
Linear Formula C3H5NH2
Quantity 10 mL
Beilstein 12,3
Specific Gravity 0.824
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 57.09
Percent Purity 99%
Physical Form Volatile Liquid
Chemical Name or Material Cyclopropylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 212-142-2

RUO – Research Use Only