missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylamine, 98+%

Catalog No. AAA1384409
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1384409 10 g
AAA1384418 50 g
AAA1384430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1384409 Supplier Thermo Scientific Chemicals Supplier No. A1384409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 765-30-0 | C3H7N | 57.096 g/mol

Cyclopropylamine) is used in the preparation of N-[4-(4-fluoro)phenyl-2-aminothiazol-5-yl]pyrimidin-2-yl-alkylamine derivatives. It is also used to prepare Pt(CPA)2(bismethylthiomethylenepropanedioate) and Pt(CPA)2(bisethylthiomethylenepropanedioate) complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropylamine) is used in the preparation of N-[4-(4-fluoro)phenyl-2-aminothiazol-5-yl]pyrimidin-2-yl-alkylamine derivatives. It is also used to prepare Pt(CPA)2(bismethylthiomethylenepropanedioate) and Pt(CPA)2(bisethylthiomethylenepropanedioate) complexes.

Solubility
Miscible with water, ethanol,ether and chloroform.

Notes
Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.

Chemical Identifiers

CAS 765-30-0
Molecular Formula C3H7N
Molecular Weight (g/mol) 57.096
MDL Number MFCD00001301
InChI Key HTJDQJBWANPRPF-UHFFFAOYSA-N
Synonym cyclopropylamine, aminocyclopropane, cyclopropyl amine, unii-8pr8xth1x1, 8pr8xth1x1, cyclopropanamin, cyclopropyamine, cyclopropylamin, cylopropylamine, cycloproplyamine
PubChem CID 69828
ChEBI CHEBI:34660
IUPAC Name cyclopropanamine
SMILES C1CC1N

Specifications

Melting Point -10°C
Density 0.824
Boiling Point 49°C to 50°C
Flash Point −25°C (−13°F)
Refractive Index 1.4206
Quantity 10 g
UN Number UN2733
Beilstein 741858
Sensitivity Hygroscopic
Solubility Information Miscible with water,ethanol,ether and chloroform.
Formula Weight 57.1
Percent Purity ≥98%
Chemical Name or Material Cyclopropylamine
Show More Show Less
Hazard Category H225-H302+H312+H332-H314-H318
Hazard Statement GHS H Statement
H225-H314-H318-H302
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P331-P363-P372-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 212-142-2
RTECSNumber GZ0685500
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only