missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylacetonitrile, 97%

Catalog No. p-7045775
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1195006 5 g
AAL1195014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1195006 Supplier Thermo Scientific Chemicals Supplier No. L1195006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6542-60-5 | C5H7N | 81.118 g/mol

Cyclopropylacetonitrile is used in the preparation of cyclopropyl-2 acetophenone and cyclopropylacetyl chloride. It acts as an organic intermediate in organic synthesis. It is also employed in the preparation of agrochemicals, pharmaceuticals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropylacetonitrile is used in the preparation of cyclopropyl-2 acetophenone and cyclopropylacetyl chloride. It acts as an organic intermediate in organic synthesis. It is also employed in the preparation of agrochemicals, pharmaceuticals and dyestuff fields.

Solubility
Miscible with water, methylene chloride and alcohols.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 6542-60-5
Molecular Formula C5H7N
Molecular Weight (g/mol) 81.118
MDL Number MFCD00041523
InChI Key FAUQRRGKJKMEIW-UHFFFAOYSA-N
Synonym cyclopropylacetonitrile, cyclopropaneacetonitrile, 2-cyclopropylethanenitrile, cyclopropyl acetonitrile, cyclopropyl-acetonitrile, acmc-20albr, cyanomethyl cyclopropane, ksc352m6t
PubChem CID 643447
IUPAC Name 2-cyclopropylacetonitrile
SMILES C1CC1CC#N

Specifications

Density 0.89
Boiling Point 142°C to 144°C
Flash Point 43°C (109°F)
Odor Strong
Refractive Index 1.4245
Quantity 5 g
UN Number UN3275
Beilstein 2036964
Solubility Information Miscible with water,methylene chloride and alcohols.
Formula Weight 81.12
Percent Purity 97%
Chemical Name or Material Cyclopropylacetonitrile
Show More Show Less
Hazard Category H226-H301+H311+H331-H341
Hazard Statement GHS H Statement
H300-H311-H331-H226-H341
Fatal if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Flammable liquid and vapor.
Suspected of causing genetic defects.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P281-P301+P310-P303+P361+P353-P304+P340-P308+P313-P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, TOXIC, FLAMMABLE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only