missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl(trimethylsilyl)acetylene, 97%

Catalog No. p-7035760
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5348706 5 g
AAH5348714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5348706 Supplier Thermo Scientific Chemicals Supplier No. H5348706
Only null left

CAS: 81166-84-9 | C8H14Si | 138.29 g/mol

Cyclopropyl(trimethylsilyl)acetylene is used in several addition reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropyl(trimethylsilyl)acetylene is used in several addition reactions.

Solubility
Sparingly Soluble (0.069 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 81166-84-9
Molecular Formula C8H14Si
Molecular Weight (g/mol) 138.29
MDL Number MFCD00671355
InChI Key IYQSVLJDYLPMCX-UHFFFAOYSA-N
Synonym cyclopropyl trimethylsilyl acetylene, cyclopropylethynyl trimethylsilane, 2-cyclopropylethynyl trimethylsilane, cyclopropylethynyl trimethyl silane, silane, cyclopropylethynyl trimethyl, cyclopropane, 2-trimethylsilyl ethynyl, silane, cyclopropylethynyl trimethyl-9ci, timtec-bb sbb009013, trimethylsilylethynylcyclopropane, cyclopropylethynyl-trimethylsilane
PubChem CID 2758018
SMILES C[Si](C)(C)C#CC1CC1

Specifications

Boiling Point 83°C (80mmHg)
Quantity 5 g
UN Number UN1993
Flash Point 25°C (77°F)
Solubility Information Sparingly Soluble (0.069g/L) (25°C).
IUPAC Name 2-cyclopropylethynyl(trimethyl)silane
Formula Weight 138.28
Percent Purity 97%
Refractive Index 1.454
Chemical Name or Material Cyclopropyl(trimethylsilyl)acetylene
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only