missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl Phenyl Ketone 98.0+%, TCI America™

Catalog No. C098725ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C098725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C098725ML Supplier TCI America Supplier No. C098725ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3481-02-5
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00001294
InChI Key PJRHFTYXYCVOSJ-UHFFFAOYSA-N
Synonym cyclopropyl phenyl ketone, cyclopropyl phenyl methanone, phenyl cyclopropyl ketone, benzoylcyclopropane, methanone, cyclopropylphenyl, cyclopropylphenylketone, ketone, cyclopropyl phenyl, cyclopropyl-phenylmethanone, acmc-2097dl, cyclopropyl-phenyl-methanone
PubChem CID 77029
IUPAC Name cyclopropyl(phenyl)methanone
SMILES C1CC1C(=O)C2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 248°C
Quantity 25 mL
Formula Weight 146.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopropyl Phenyl Ketone