missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl phenyl ketone, 97%

Catalog No. AAL0221822
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0221822 100 g
AAL0221814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0221822 Supplier Thermo Scientific Chemicals Supplier No. L0221822

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3481-02-5 | C10H10O | 146.19 g/mol

Cyclopropyl phenyl ketone is used to prepare α-cyclopropylstyrene by the Wittig reaction in dimethyl sulfoxide. It is also used as starting reagent during the (Z)-titanium enolate formation from cyclopropyl ketones via TiCl4-n-Bu4NI-induced ring opening reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropyl phenyl ketone is used to prepare α-cyclopropylstyrene by the Wittig reaction in dimethyl sulfoxide. It is also used as starting reagent during the (Z)-titanium enolate formation from cyclopropyl ketones via TiCl4-n-Bu4NI-induced ring opening reaction.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 3481-02-5
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00001294
InChI Key PJRHFTYXYCVOSJ-UHFFFAOYSA-N
Synonym cyclopropyl phenyl ketone, cyclopropyl phenyl methanone, phenyl cyclopropyl ketone, benzoylcyclopropane, methanone, cyclopropylphenyl, cyclopropylphenylketone, ketone, cyclopropyl phenyl, cyclopropyl-phenylmethanone, acmc-2097dl, cyclopropyl-phenyl-methanone
PubChem CID 77029
IUPAC Name cyclopropyl(phenyl)methanone
SMILES C1CC1C(=O)C2=CC=CC=C2

Specifications

Density 1.06
Boiling Point 248°C
Flash Point 90°C (194°F)
Refractive Index 1.5534
Quantity 100 g
Beilstein 1860145
Solubility Information Insoluble in water.
Formula Weight 146.19
Percent Purity 97%
Chemical Name or Material Cyclopropyl phenyl ketone
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 222-458-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only