Learn More
Cyclopropanemethylamine hydrochloride, 98%
CAS: 7252-53-1 | C4H9ClN | 106.57 g/mol
$81.56 - $245.38
Chemical Identifiers
| CAS | 7252-53-1 |
|---|---|
| Molecular Formula | C4H9ClN |
| Molecular Weight (g/mol) | 106.57 |
| MDL Number | MFCD00012544 |
| InChI Key | MQCZBGGAZQXTLH-UHFFFAOYSA-N |
| Synonym | cyclopropanemethylamine hydrochloride, cyclopropylmethanamine hydrochloride, cyclopropylmethyl ammonium chloride, cyclopropylmethylamine hydrochloride, aminomethylcyclopropane hydrochloride, 1-cyclopropylmethanamine hydrochloride, aminomethyl cyclopropane hydrochloride, cyclopropylmethanaminium chloride, cyclopropylmethylamin hydrochloride, cyclopropylmethanamine-hydrochloride |
| PubChem CID | 81671 |
| IUPAC Name | cyclopropylmethanamine;hydrochloride |
| SMILES | [Cl].NCC1CC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1335303
|
Thermo Scientific Chemicals
L1335303 |
1 g |
Each for $81.56
|
|
|||||
|
AAL1335306
|
Thermo Scientific Chemicals
L1335306 |
5 g |
Each for $245.38
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7252-53-1 | |
| 106.57 | |
| MQCZBGGAZQXTLH-UHFFFAOYSA-N | |
| 81671 | |
| [Cl].NCC1CC1 |
| C4H9ClN | |
| MFCD00012544 | |
| cyclopropanemethylamine hydrochloride, cyclopropylmethanamine hydrochloride, cyclopropylmethyl ammonium chloride, cyclopropylmethylamine hydrochloride, aminomethylcyclopropane hydrochloride, 1-cyclopropylmethanamine hydrochloride, aminomethyl cyclopropane hydrochloride, cyclopropylmethanaminium chloride, cyclopropylmethylamin hydrochloride, cyclopropylmethanamine-hydrochloride | |
| cyclopropylmethanamine;hydrochloride |
Specifications
| 7252-53-1 | |
| C4H9ClN | |
| 1 g | |
| Hygroscopic | |
| MQCZBGGAZQXTLH-UHFFFAOYSA-N | |
| cyclopropylmethanamine;hydrochloride | |
| 81671 | |
| 98% |
| 202°C to 205°C | |
| MFCD00012544 | |
| 3906936 | |
| cyclopropanemethylamine hydrochloride, cyclopropylmethanamine hydrochloride, cyclopropylmethyl ammonium chloride, cyclopropylmethylamine hydrochloride, aminomethylcyclopropane hydrochloride, 1-cyclopropylmethanamine hydrochloride, aminomethyl cyclopropane hydrochloride, cyclopropylmethanaminium chloride, cyclopropylmethylamin hydrochloride, cyclopropylmethanamine-hydrochloride | |
| [Cl].NCC1CC1 | |
| 106.57 | |
| 107.58 | |
| Cyclopropanemethylamine hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 230-668-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only