missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanemethylamine hydrochloride, 98%

Catalog No. AAL1335303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1335303 1 g
AAL1335306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1335303 Supplier Thermo Scientific Chemicals Supplier No. L1335303

CAS: 7252-53-1 | C4H9ClN | 106.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7252-53-1
Molecular Formula C4H9ClN
Molecular Weight (g/mol) 106.57
MDL Number MFCD00012544
InChI Key MQCZBGGAZQXTLH-UHFFFAOYSA-N
Synonym cyclopropanemethylamine hydrochloride, cyclopropylmethanamine hydrochloride, cyclopropylmethyl ammonium chloride, cyclopropylmethylamine hydrochloride, aminomethylcyclopropane hydrochloride, 1-cyclopropylmethanamine hydrochloride, aminomethyl cyclopropane hydrochloride, cyclopropylmethanaminium chloride, cyclopropylmethylamin hydrochloride, cyclopropylmethanamine-hydrochloride
PubChem CID 81671
IUPAC Name cyclopropylmethanamine;hydrochloride
SMILES [Cl].NCC1CC1

Specifications

Melting Point 202°C to 205°C
Quantity 1 g
Beilstein 3906936
Sensitivity Hygroscopic
Formula Weight 107.58
Percent Purity 98%
Chemical Name or Material Cyclopropanemethylamine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 230-668-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only