missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanemethylamine, 97%

Catalog No. AAA1394614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1394614 25 g
AAA1394603 1 g
AAA1394606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1394614 Supplier Thermo Scientific Chemicals Supplier No. A1394614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2516-47-4 | C4H9N | 71.123 g/mol

Cyclopropanemethylamine used in studies of potential inhibitors of glycine cleavage system of brain and liver mitochondria. It is used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropanemethylamine used in studies of potential inhibitors of glycine cleavage system of brain and liver mitochondria. It is used in chemical synthesis.

Solubility
Fully miscible in water.

Notes
Air sensitive. Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, air , carbon dioxide.

Chemical Identifiers

CAS 2516-47-4
Molecular Formula C4H9N
Molecular Weight (g/mol) 71.123
MDL Number MFCD00037147
InChI Key IGSKHXTUVXSOMB-UHFFFAOYSA-N
Synonym cyclopropylmethylamine, cyclopropanemethylamine, cyclopropanemethanamine, aminomethylcyclopropane, aminomethyl cyclopropane, 1-cyclopropylmethanamine, cyclopropane methyl amine, cyclopropylmethylamin, cyclopropanmethylamine
PubChem CID 75646
IUPAC Name cyclopropylmethanamine
SMILES C1CC1CN

Specifications

Density 0.82
Boiling Point 85°C to 86°C
Flash Point 2°C (36°F)
Odor Amine-like
Refractive Index 1.433
Quantity 25 g
UN Number UN2733
Beilstein 773674
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
Formula Weight 71.12
Percent Purity ≥98%
Chemical Name or Material Cyclopropanemethylamine
Show More Show Less
Hazard Category H225-H314
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 219-737-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only