missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanecarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7124389
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C038725ML 25 mL
C0387100ML 100 mL
C0387500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C038725ML Supplier TCI America Supplier No. C038725ML
Only null left

Chemical Identifiers

CAS 1759-53-1
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00001287
InChI Key YMGUBTXCNDTFJI-UHFFFAOYSA-N
Synonym carboxycyclopropane, cyclopropylcarboxylic acid, cyclopropionic acid, cyclopropane carboxylic acid, cyclopropanecarboxylicacid, cpc-acid, cyclopropyl carboxylic acid, trimethylenecarboxylic acid, cyclopropane carboylic acid, cyclopropane-carboxylic acid
PubChem CID 15655
ChEBI CHEBI:23500
IUPAC Name cyclopropanecarboxylic acid
SMILES C1CC1C(=O)O

Specifications

Color Yellow
Boiling Point 183°C
Quantity 25 mL
UN Number 3265
Formula Weight 86.09
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Cyclopropanecarboxylic Acid
EINECSNumber (3)-4419
RTECSNumber GZ1100000
TSCA Yes