missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanecarboxylic acid, 98%

Catalog No. p-7021749
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1421518 50 g
AAA1421530 250 g
AAA142150B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1421518 Supplier Thermo Scientific Chemicals Supplier No. A1421518
Only null left

CAS: 1759-53-1 | C4H6O2 | 86.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1759-53-1
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00001287
InChI Key YMGUBTXCNDTFJI-UHFFFAOYSA-N
Synonym carboxycyclopropane, cyclopropylcarboxylic acid, cyclopropionic acid, cyclopropane carboxylic acid, cyclopropanecarboxylicacid, cpc-acid, cyclopropyl carboxylic acid, trimethylenecarboxylic acid, cyclopropane carboylic acid, cyclopropane-carboxylic acid
PubChem CID 15655
ChEBI CHEBI:23500
IUPAC Name cyclopropanecarboxylic acid
SMILES C1CC1C(=O)O

Specifications

Melting Point 5°C
Density 1.086
Boiling Point 182°C to 184°C
Flash Point 71°C (159°F)
Refractive Index 1.439
Quantity 50 g
UN Number UN3265
Beilstein 969839
Formula Weight 86.09
Percent Purity 98%
Chemical Name or Material Cyclopropanecarboxylic acid
Show More Show Less
Hazard Category H227-H302-H314
Hazard Statement GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 217-162-5
RTECSNumber GZ1100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only