missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropanecarboxylic acid, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 17.0°C to 19.0°C |
| Color | Colorless to Yellow |
| Density | 1.0850g/mL |
| Boiling Point | 182.0°C to 184.0°C |
| Flash Point | 71°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.4370 to 1.4390 |
| Linear Formula | C3H5CO2H |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Harmful if swallowed. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for |
| EINECSNumber | 217-162-5 |
| RTECSNumber | GZ1100000 |
| TSCA | TSCA |
RUO – Research Use Only