missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanecarboxamidine Hydrochloride 98.0+%, TCI America™

Catalog No. C23441G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23441G 1 g
C23445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23441G Supplier TCI America Supplier No. C23441G

Chemical Identifiers

CAS 57297-29-7
Molecular Formula C4H9ClN2
Molecular Weight (g/mol) 120.58
MDL Number MFCD00053010
InChI Key JRYOZJIRAVZGMV-UHFFFAOYSA-N
Synonym cyclopropanecarboximidamide hydrochloride, cyclopropane-1-carboximidamide hydrochloride, cyclopropanecarboxamidine hydrochloride, cyclopropylcarbamidine hydrochloride, cyclopropanecarboxamidine hcl, cyclopropyl carbamidine hydrochloride, clcyopropylcarbamidine hydrochloride, cyclopropylcarboxamidine hydrochloride, cyclopropyl carbamidine hcl, cyclopropanecarboximidamide, monohydrochloride
PubChem CID 2781916
IUPAC Name hydrogen cyclopropanecarboximidamide chloride
SMILES [H+].[Cl-].NC(=N)C1CC1

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 1 g
Formula Weight 120.58
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Cyclopropanecarboxamidine Hydrochloride