missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanecarboxamide 98.0+%, TCI America™

Catalog No. C201025G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C201025G 25 g
C2010250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C201025G Supplier TCI America Supplier No. C201025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6228-73-5
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.106
InChI Key AIMMVWOEOZMVMS-UHFFFAOYSA-N
Synonym cyclopropylcarboxamide, carbamoylcyclopropane, cyclopropyl carboxamide, seed one, cyclopropanecarboxylic acid amide, cyclopropanamide, cyclopropancarboxamide, carbamoyl cyclopropane, cyclopropane carboxamide, pubchem21401
PubChem CID 80376
ChEBI CHEBI:51457
IUPAC Name cyclopropanecarboxamide
SMILES C1CC1C(=O)N

Specifications

Melting Point 122°C
Quantity 25 g
Formula Weight 85.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Cyclopropanecarboxamide