Learn More
Cyclopentylmagnesium chloride, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific™
Chemical Identifiers
| CAS | 32916-51-1 |
|---|---|
| Molecular Formula | C5H9ClMg |
| Molecular Weight (g/mol) | 128.88 |
| MDL Number | MFCD00001357 |
| InChI Key | KQIVBIRNYZIHNE-UHFFFAOYSA-M |
| Synonym | cyclopentylmagnesium chloride, cyclopentyl magnesium chloride, cyclopentylmagnesium chloride solution, cylopentylmagnesium chloride, cyclopentylmagnesium cloride, magnesium,chlorocyclopentyl, cylopentyl magnesium chloride, cyclopentyl-magnesium chloride, grogeabzckqtjw-uhfffaoysa-m, chloro cyclopentyl magnesium |
| PubChem CID | 3599845 |
| IUPAC Name | magnesium;cyclopentane;chloride |
| SMILES | [Mg++].[Cl-].C1CC[CH-]C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC208461000
|
Thermo Scientific Chemicals
208461000 |
100mL | AcroSeal™ Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC208468000
|
Thermo Scientific Chemicals
208468000 |
800mL | AcroSeal™ Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 32916-51-1 | |
| 128.88 | |
| KQIVBIRNYZIHNE-UHFFFAOYSA-M | |
| 3599845 | |
| [Mg++].[Cl-].C1CC[CH-]C1 |
| C5H9ClMg | |
| MFCD00001357 | |
| cyclopentylmagnesium chloride, cyclopentyl magnesium chloride, cyclopentylmagnesium chloride solution, cylopentylmagnesium chloride, cyclopentylmagnesium cloride, magnesium,chlorocyclopentyl, cylopentyl magnesium chloride, cyclopentyl-magnesium chloride, grogeabzckqtjw-uhfffaoysa-m, chloro cyclopentyl magnesium | |
| magnesium;cyclopentane;chloride |
Specifications
| 32916-51-1,60-29-7 | |
| −40°C | |
| C5H9ClMg | |
| MFCD00001357 | |
| 0.86 | |
| KQIVBIRNYZIHNE-UHFFFAOYSA-M | |
| magnesium;cyclopentane;chloride | |
| 3599845 | |
| Cyclopentylmagnesium chloride, 2M solution in diethyl ether |
| 0.8600g/mL | |
| AcroSeal™ Glass Bottle | |
| C5H9MgCl | |
| 100mL | |
| cyclopentylmagnesium chloride, cyclopentyl magnesium chloride, cyclopentylmagnesium chloride solution, cylopentylmagnesium chloride, cyclopentylmagnesium cloride, magnesium,chlorocyclopentyl, cylopentyl magnesium chloride, cyclopentyl-magnesium chloride, grogeabzckqtjw-uhfffaoysa-m, chloro cyclopentyl magnesium | |
| [Mg++].[Cl-].C1CC[CH-]C1 | |
| 128.88 | |
| 128.88 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
In contact with water releases flammable gas.
Extremely flammable liquid and vapor.
May form explosive
GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
RUO â Research Use Only