missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopentylboronic Acid (contains varying amounts of Anhydride), TCI America™
$46.60 - $46.60
Chemical Identifiers
| CAS | 63076-51-7 |
|---|---|
| Molecular Formula | C5H11BO2 |
| MDL Number | MFCD01074541 |
| InChI Key | VTTDFSNKIMAQTB-UHFFFAOYSA-N |
| PubChem CID | 2734327 |
| IUPAC Name | cyclopentylboronic acid |
Chemical Identifiers
| 63076-51-7 | |
| MFCD01074541 | |
| 2734327 |
| C5H11BO2 | |
| VTTDFSNKIMAQTB-UHFFFAOYSA-N | |
| cyclopentylboronic acid |
Specifications
| 63076-51-7 | |
| C5H11BO2 | |
| 1 g | |
| VTTDFSNKIMAQTB-UHFFFAOYSA-N | |
| cyclopentylboronic acid | |
| 2734327 | |
| Crystalline Powder |
| 126°C | |
| MFCD01074541 | |
| cyclopentaneboronic acid, boronic acid, cyclopentyl, cylopentyl boronic acid, cyclopentylboronicacid, pubchem7792, acmc-209ncu, --paclitaxel;paclitaxel, boronic acid, b-cyclopentyl, cyclopentylboronic acid | |
| B(C1CCCC1)(O)O | |
| 113.951 | |
| 113.95 | |
| Cyclopentylboronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
TSCA : No