missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylamine 99.0+%, TCI America™

Catalog No. p-7124664
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
C088725ML 25 mL
C0887500ML 500 mL
2 options

Catalog No. C088725ML

Supplier: TCI America C088725ML

Only null left

Chemical Identifiers

CAS 1003-03-8
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number MFCD00001380
InChI Key NISGSNTVMOOSJQ-UHFFFAOYSA-N
Synonym cyclopentylamine, aminocyclopentane, amino cyclopentane, unii-4259vry3gn, 1cb, cylcopentylamine, cyclopentyl amine, cyclopentyl-amine, n-cyclopentylamine, cyclo-pentyl amine
PubChem CID 2906
IUPAC Name cyclopentanamine
SMILES NC1CCCC1

Specifications

Melting Point -86°C
Color Colorless
Boiling Point 107°C
Quantity 25 mL
UN Number 2733
Formula Weight 85.15
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Cyclopentylamine
RTECSNumber GY8452000
TSCA Yes